The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173799.3 | NP_776160.2 | 2 - 3 | 467 | 73 |
XM_011512538.1 | XP_011510840.1 | 2 - 3 | 455 | 73 | |
GenBank mRNA | AK097192.1 | - | 2 - 3 | 425 | 73 |
AL833175.1 | - | 3 - 4 | 3207 | 73 | |
BC101288.1 | - | 2 - 3 | 424 | 73 | |
BC101289.1 | - | 2 - 3 | 424 | 73 | |
BC101290.1 | - | 2 - 3 | 424 | 73 | |
BC101291.1 | - | 2 - 3 | 424 | 73 | |
BX640915.1 | - | 1 - 2 | 331 | 73 | |
EU675310.1 | - | 2 - 3 | 467 | 73 |