Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IFT20
Entrez Gene ID
90410
Gene Name
intraflagellar transport 20
Gene Aliases
-
Chromosome Location
Chr.17: 28328325 - 28335489 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001267774.1 NP_001254703.1 2 - 3 388 104
GenBank mRNA AK297769.1 - 2 - 3 354 104
AK309232.1 - 2 - 3 377 104
AL713094.1 - 2 - 3 389 104
AY224601.1 - 2 - 3 337 104

View Full Product Details