The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001272065.1 | NP_001258994.1 | 4 - 5 | 564 | 139 |
NM_001272066.1 | NP_001258995.1 | 4 - 5 | 551 | 139 | |
NM_001272067.1 | NP_001258996.1 | 4 - 5 | 746 | 139 | |
NM_001272068.1 | NP_001258997.1 | 5 - 6 | 783 | 139 | |
NM_001272082.1 | NP_001259011.1 | 2 - 3 | 440 | 139 | |
NM_001272083.1 | NP_001259012.1 | 2 - 3 | 440 | 139 | |
NM_016479.4 | NP_057563.3 | 4 - 5 | 585 | 139 | |
XM_005265205.1 | XP_005265262.1 | 5 - 6 | 833 | 139 | |
XM_006713188.2 | XP_006713251.1 | 4 - 5 | 486 | 139 | |
XM_011533796.1 | XP_011532098.1 | 5 - 6 | 575 | 139 | |
GenBank mRNA | AB097007.1 | - | 2 - 3 | 204 | 139 |
AF520698.1 | - | 4 - 5 | 564 | 139 | |
AK056328.1 | - | 4 - 5 | 727 | 139 | |
AK075441.1 | - | 4 - 5 | 500 | 139 | |
AK124652.1 | - | 5 - 6 | 783 | 139 | |
AK298799.1 | - | 2 - 3 | 265 | 139 | |
AK298842.1 | - | 2 - 3 | 215 | 139 | |
AK301191.1 | - | 2 - 3 | 499 | 139 | |
AL117413.1 | - | 2 - 3 | 211 | 139 | |
AL832568.1 | - | 2 - 3 | 247 | 139 | |
BC001463.2 | - | 2 - 3 | 255 | 139 | |
BC020279.2 | - | 2 - 3 | 205 | 139 | |
BX336842.2 | - | 4 - 5 | 468 | 139 | |
BX344125.2 | - | 4 - 5 | 511 | 139 | |
BX379507.2 | - | 4 - 5 | 456 | 140 | |
DA046900.1 | - | 2 - 3 | 440 | 139 |