The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000967.3 | NP_000958.1 | 6 - 7 | 910 | 68 |
NM_001033853.1 | NP_001029025.1 | 6 - 7 | 763 | 68 | |
GenBank mRNA | AB062291.1 | - | 5 - 6 | 847 | 68 |
AB266619.1 | - | 5 - 6 | 686 | 68 | |
AK092695.1 | - | 6 - 7 | 721 | 68 | |
AK315693.1 | - | 6 - 7 | 868 | 68 | |
BC002408.2 | - | 5 - 6 | 845 | 68 | |
BC004323.1 | - | 4 - 5 | 509 | 68 | |
BC006483.1 | - | 6 - 7 | 855 | 68 | |
BC008003.1 | - | 5 - 6 | 847 | 68 | |
BC008492.1 | - | 6 - 7 | 868 | 68 | |
BC012146.1 | - | 6 - 7 | 855 | 68 | |
BC012786.2 | - | 6 - 7 | 855 | 68 | |
BC013674.1 | - | 5 - 6 | 851 | 68 | |
BC013861.1 | - | 5 - 6 | 851 | 68 | |
BC014017.2 | - | 5 - 6 | 850 | 68 | |
BC015032.1 | - | 5 - 6 | 851 | 68 | |
BC015767.1 | - | 5 - 6 | 851 | 68 | |
BC022790.1 | - | 5 - 6 | 756 | 68 | |
BC033296.1 | - | 5 - 6 | 848 | 68 | |
BC036582.1 | - | 6 - 7 | 1687 | 68 | |
BC063662.1 | - | 5 - 6 | 855 | 68 | |
BC088373.1 | - | 6 - 7 | 865 | 68 | |
BC107711.1 | - | 6 - 7 | 872 | 68 | |
CR456566.1 | - | 5 - 6 | 854 | 68 | |
M90054.1 | - | 5 - 6 | 828 | 68 | |
X73460.1 | - | 5 - 6 | 848 | 68 |