Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IRX6
Entrez Gene ID
79190
Gene Name
iroquois homeobox 6
Gene Aliases
IRX-3, IRX7, IRXB3
Chromosome Location
Chr.16: 55323962 - 55330760 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005256137.2 XP_005256194.1 5 - 6 2467 152
GenBank mRNA AY335937.1 - 5 - 6 1870 152

View Full Product Details