Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYLK4
Entrez Gene ID
340156
Gene Name
myosin light chain kinase family member 4
Gene Aliases
SgK085
Chromosome Location
Chr.6: 2663629 - 2770330 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012418.3 NP_001012418.2 8 - 9 1055 154
XM_005249078.3 XP_005249135.2 8 - 9 1020 154
XM_005249079.2 XP_005249136.1 8 - 9 913 154
XM_006715082.3 XP_006715145.1 7 - 8 2350 154
XM_011514567.2 XP_011512869.1 8 - 9 1143 154
XM_017010819.1 XP_016866308.1 8 - 9 952 154
GenBank mRNA BC132831.1 - 7 - 8 821 154
BC132833.1 - 7 - 8 821 154
BX647582.1 - 8 - 9 1055 154

View Full Product Details