Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GBP6
Entrez Gene ID
163351
Gene Name
guanylate binding protein family member 6
Gene Aliases
-
Chromosome Location
Chr.1: 89363877 - 89388160 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001320257.1 NP_001307186.1 5 - 6 1084 152
NM_198460.2 NP_940862.2 7 - 8 1425 152
XM_011540835.2 XP_011539137.1 7 - 8 1254 152
GenBank mRNA AK131329.1 - 2 - 3 307 152
AK131356.1 - 7 - 8 1409 152
AK290303.1 - 7 - 8 1245 152
AK299408.1 - 6 - 7 1060 152
AK299466.1 - 5 - 6 901 152
AK303987.1 - 7 - 8 1169 152
AK315834.1 - 7 - 8 1243 152
BC131713.1 - 7 - 8 1203 152
BX537949.1 - 7 - 8 1244 152
BX647907.1 - 7 - 8 1242 152

View Full Product Details