Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GBP6
Entrez Gene ID
163351
Gene Name
guanylate binding protein family member 6
Gene Aliases
-
Chromosome Location
Chr.1: 89363877 - 89388160 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001320257.1 NP_001307186.1 7 - 8 1401 108
NM_198460.2 NP_940862.2 9 - 10 1742 108
XM_011540835.2 XP_011539137.1 9 - 10 1571 108
GenBank mRNA AK131329.1 - 4 - 5 624 108
AK131356.1 - 9 - 10 1726 108
AK299408.1 - 8 - 9 1377 108
AK299466.1 - 7 - 8 1218 108
AK303987.1 - 9 - 10 1486 108
AK315834.1 - 9 - 10 1560 108
BC131713.1 - 9 - 10 1520 108
BX537949.1 - 9 - 10 1561 108
BX647907.1 - 9 - 10 1559 108

View Full Product Details