Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IFT20
Entrez Gene ID
90410
Gene Name
intraflagellar transport 20
Gene Aliases
-
Chromosome Location
Chr.17: 28328325 - 28335489 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001267774.1 NP_001254703.1 5 - 6 707 139
NM_001267775.1 NP_001254704.1 5 - 6 506 139
NM_001267776.1 NP_001254705.1 4 - 5 470 139
NM_174887.3 NP_777547.1 5 - 6 588 139
GenBank mRNA AY224601.1 - 5 - 6 656 139
BC002640.2 - 5 - 6 568 139
BC038094.1 - 4 - 5 436 139

View Full Product Details