Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C5orf47
Entrez Gene ID
133491
Gene Name
chromosome 5 open reading frame 47
Gene Aliases
-
Chromosome Location
Chr.5: 173989159 - 174007524 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144954.1 NP_001138426.1 2 - 3 513 128
XM_017009028.1 XP_016864517.1 2 - 3 507 128
GenBank mRNA BC034971.1 - 2 - 3 479 128
BC047225.1 - 2 - 3 468 128
BC053953.1 - 2 - 3 468 128
BC057842.1 - 2 - 3 454 128
BC066964.1 - 2 - 3 475 128
BC092489.1 - 2 - 3 502 128

View Full Product Details