The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001144954.1 | NP_001138426.1 | 4 - 5 | 651 | 99 |
GenBank mRNA | BC034971.1 | - | 4 - 5 | 617 | 99 |
BC047225.1 | - | 4 - 5 | 606 | 99 | |
BC053953.1 | - | 4 - 5 | 606 | 99 | |
BC057842.1 | - | 4 - 5 | 592 | 99 | |
BC066964.1 | - | 4 - 5 | 613 | 99 | |
BC092489.1 | - | 4 - 5 | 640 | 99 |