The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001031677.3 | NP_001026847.1 | 3 - 4 | 718 | 163 |
NM_130781.3 | NP_570137.2 | 4 - 5 | 632 | 163 | |
NR_109789.1 | 4 - 5 | 632 | 163 | ||
NR_109790.1 | 3 - 4 | 718 | 163 | ||
GenBank mRNA | AF087904.1 | - | 3 - 4 | 436 | 163 |
AF258583.1 | - | 3 - 4 | 462 | 163 | |
AK021761.1 | - | 2 - 3 | 749 | 163 | |
AK308155.1 | - | 4 - 5 | 587 | 163 | |
AL833898.1 | - | 3 - 4 | 260 | 163 | |
BC010006.2 | - | 4 - 5 | 555 | 163 | |
BC015534.1 | - | 4 - 5 | 594 | 163 | |
BC021263.2 | - | 3 - 4 | 678 | 163 | |
BT007268.1 | - | 3 - 4 | 262 | 163 |