Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
XG
Entrez Gene ID
7499
Gene Name
Xg blood group
Gene Aliases
PBDX
Chromosome Location
Chr.X: 2751108 - 2816500 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001141919.1 NP_001135391.1 3 - 4 344 146
NM_001141920.1 NP_001135392.1 3 - 4 344 146
NM_175569.2 NP_780778.1 3 - 4 344 146
XM_005274587.3 XP_005274644.1 3 - 4 1272 146
XM_011545575.2 XP_011543877.1 3 - 4 1288 146
GenBank mRNA AF380356.1 - 3 - 4 322 146
AF534880.1 - 3 - 4 318 146
AK304164.1 - 3 - 4 344 146
BC100764.2 - 4 - 5 309 146
BC100765.2 - 3 - 4 206 146
BC100766.2 - 3 - 4 207 146
BC100767.1 - 3 - 4 206 146

View Full Product Details