The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001141919.1 | NP_001135391.1 | 5 - 6 | 473 | 105 |
NM_175569.2 | NP_780778.1 | 5 - 6 | 473 | 105 | |
GenBank mRNA | AF380356.1 | - | 5 - 6 | 451 | 105 |
AF534880.1 | - | 5 - 6 | 447 | 105 | |
AK304164.1 | - | 5 - 6 | 473 | 105 | |
BC100765.2 | - | 5 - 6 | 335 | 105 | |
BC100766.2 | - | 5 - 6 | 336 | 105 |