Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SMYD3
Entrez Gene ID
64754
Gene Name
SET and MYND domain containing 3
Gene Aliases
KMT3E, ZMYND1, ZNFN3A1, bA74P14.1
Chromosome Location
Chr.1: 245749340 - 246507342 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001167740.1 NP_001161212.1 3 - 4 467 163
NM_022743.2 NP_073580.1 3 - 4 295 163
XM_011544253.1 XP_011542555.1 3 - 4 443 163
GenBank mRNA AB057595.1 - 3 - 4 437 163
AK024733.1 - 3 - 4 292 163
AK289605.1 - 3 - 4 443 163
BC107725.1 - 3 - 4 441 163

View Full Product Details