The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001167740.1 | NP_001161212.1 | 9 - 10 | 1030 | 155 |
NM_022743.2 | NP_073580.1 | 9 - 10 | 858 | 155 | |
XM_011544253.1 | XP_011542555.1 | 8 - 9 | 895 | 155 | |
XM_011544254.2 | XP_011542556.1 | 5 - 6 | 1002 | 155 | |
XM_011544257.1 | XP_011542559.1 | 5 - 6 | 494 | 155 | |
XM_011544258.2 | XP_011542560.1 | 6 - 7 | 851 | 155 | |
XM_011544259.2 | XP_011542561.1 | 5 - 6 | 717 | 155 | |
XM_011544260.1 | XP_011542562.1 | 5 - 6 | 534 | 155 | |
XM_011544261.2 | XP_011542563.1 | 5 - 6 | 459 | 155 | |
XM_017002094.1 | XP_016857583.1 | 5 - 6 | 4674 | 155 | |
XM_017002095.1 | XP_016857584.1 | 5 - 6 | 405 | 155 | |
GenBank mRNA | AB057595.1 | - | 9 - 10 | 1000 | 155 |
AK023594.1 | - | 4 - 5 | 1260 | 155 | |
AK024733.1 | - | 9 - 10 | 855 | 155 | |
AK289605.1 | - | 9 - 10 | 1006 | 155 | |
AY186742.1 | - | 6 - 7 | 769 | 155 | |
BC017079.1 | - | 5 - 6 | 482 | 155 | |
BC031010.1 | - | 9 - 10 | 945 | 155 |