Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C2orf78
Entrez Gene ID
388960
Gene Name
chromosome 2 open reading frame 78
Gene Aliases
COG5373, hCG1989538
Chromosome Location
Chr.2: 73784189 - 73817148 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080474.1 NP_001073943.1 2 - 3 969 158
XM_005264302.3 XP_005264359.1 2 - 3 914 158
GenBank mRNA AK125975.1 - 2 - 3 969 158

View Full Product Details