Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MYO10
Entrez Gene ID
4651
Gene Name
myosin X
Gene Aliases
-
Chromosome Location
Chr.5: 16661907 - 16936276 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012334.2 NP_036466.2 12 - 13 1792 144
XM_006714475.2 XP_006714538.1 12 - 13 1568 144
GenBank mRNA AB018342.2 - 12 - 13 1779 144
AF132021.1 - 12 - 13 1404 144
AF184153.1 - 3 - 4 286 144
AF234532.1 - 12 - 13 1546 144
AF247457.2 - 12 - 13 1477 144
AK024719.1 - 3 - 4 403 144
AK125246.1 - 12 - 13 1776 144
AL832428.1 - 2 - 3 249 144
BC014956.1 - 12 - 13 1788 144
BC137168.1 - 12 - 13 1501 144
BC150285.1 - 12 - 13 1779 144

View Full Product Details