Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MYO10
Entrez Gene ID
4651
Gene Name
myosin X
Gene Aliases
-
Chromosome Location
Chr.5: 16661907 - 16936276 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012334.2 NP_036466.2 31 - 32 4664 132
XM_005248306.4 XP_005248363.1 13 - 14 3558 132
XM_005248307.2 XP_005248364.1 13 - 14 3034 132
XM_006714475.2 XP_006714538.1 30 - 31 4371 132
XM_011514046.2 XP_011512348.1 13 - 14 2737 132
GenBank mRNA AB018342.2 - 31 - 32 4651 132
AF132021.1 - 31 - 32 4276 132
AF247457.2 - 31 - 32 4349 132
AK074717.1 - 7 - 8 1010 132
AK295630.1 - 6 - 7 912 132
AK295736.1 - 13 - 14 2519 132
AK303459.1 - 1 - 2 258 132
AK316138.1 - 13 - 14 2692 132
AL832428.1 - 21 - 22 3118 132
BC137168.1 - 31 - 32 4373 132
BC150285.1 - 31 - 32 4651 132

View Full Product Details