Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARNTL
Entrez Gene ID
406
Gene Name
aryl hydrocarbon receptor nuclear translocator like
Gene Aliases
BMAL1, BMAL1c, JAP3, MOP3, PASD3, TIC, bHLHe5
Chromosome Location
Chr.11: 13276613 - 13387268 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030272.2 NP_001025443.1 17 - 18 2025 150
NM_001030273.2 NP_001025444.1 18 - 19 2096 150
NM_001178.5 NP_001169.3 18 - 19 2080 150
NM_001297719.1 NP_001284648.1 18 - 19 2083 150
NM_001297722.1 NP_001284651.1 17 - 18 2028 150
NM_001297724.1 NP_001284653.1 18 - 19 2092 150
XM_006718233.3 XP_006718296.1 17 - 18 1985 150
XM_006718234.1 XP_006718297.1 18 - 19 2051 150
XM_011520105.2 XP_011518407.2 16 - 17 2146 150
XM_011520107.2 XP_011518409.2 16 - 17 2126 150
XM_011520108.1 XP_011518410.1 18 - 19 2091 150
XM_011520109.1 XP_011518411.1 16 - 17 1846 150
XM_011520110.1 XP_011518412.1 19 - 20 2106 150
XM_011520111.1 XP_011518413.1 17 - 18 1921 150
XM_011520112.2 XP_011518414.1 19 - 20 2091 150
XM_011520113.1 XP_011518415.1 19 - 20 2088 150
XM_017017738.1 XP_016873227.1 16 - 17 2147 150
XM_017017739.1 XP_016873228.1 16 - 17 2128 150
XM_017017740.1 XP_016873229.1 17 - 18 1982 150
XM_017017741.1 XP_016873230.1 16 - 17 1843 150
XM_017017742.1 XP_016873231.1 16 - 17 1831 150
XM_017017743.1 XP_016873232.1 16 - 17 1828 150
XM_017017744.1 XP_016873233.1 19 - 20 2103 150
XM_017017745.1 XP_016873234.1 18 - 19 2048 150
XM_017017746.1 XP_016873235.1 18 - 19 2046 150
XM_017017747.1 XP_016873236.1 17 - 18 1906 150
XM_017017748.1 XP_016873237.1 17 - 18 1960 150
GenBank mRNA AB000812.1 - 15 - 16 1664 150
AB000814.1 - 4 - 5 480 150
AF044288.1 - 18 - 19 1993 150
AK095749.1 - 18 - 19 1981 150
AK128317.1 - 3 - 4 3098 150
AK291510.1 - 17 - 18 1941 150
BC016674.1 - 15 - 16 1627 150
BC031214.1 - 18 - 19 1924 150
BC041129.1 - 18 - 19 2004 150
D89722.1 - 15 - 16 1640 150
U51627.1 - 12 - 13 1622 150
U60415.1 - 17 - 18 1927 150

View Full Product Details