The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001207042.2 | NP_001193971.1 | 3 - 4 | 426 | 173 |
NM_001536.5 | NP_001527.3 | 4 - 5 | 480 | 173 | |
NM_198318.4 | NP_938074.2 | 3 - 4 | 426 | 173 | |
NR_033397.4 | 5 - 6 | 598 | 173 | ||
XM_005258842.1 | XP_005258899.1 | 5 - 6 | 468 | 173 | |
XM_017026734.1 | XP_016882223.1 | 5 - 6 | 484 | 173 | |
XM_017026735.1 | XP_016882224.1 | 5 - 6 | 627 | 173 | |
XM_017026736.1 | XP_016882225.1 | 5 - 6 | 692 | 173 | |
GenBank mRNA | AK295973.1 | - | 5 - 6 | 515 | 173 |
AK304660.1 | - | 3 - 4 | 426 | 173 | |
AL711069.1 | - | 4 - 5 | 374 | 173 | |
BC019268.1 | - | 3 - 4 | 296 | 173 | |
BC109282.1 | - | 3 - 4 | 283 | 173 | |
BC109283.2 | - | 3 - 4 | 283 | 173 | |
BX352789.2 | - | 3 - 4 | 293 | 173 | |
CR407608.1 | - | 3 - 4 | 279 | 173 | |
DC421815.1 | - | 3 - 4 | 426 | 173 |