The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001098479.1 | NP_001091949.1 | 6 - 7 | 1161 | 107 |
XM_017010810.1 | XP_016866299.1 | 6 - 7 | 1067 | 107 | |
XM_017010812.1 | XP_016866301.1 | 6 - 7 | 1067 | 107 | |
XM_017010813.1 | XP_016866302.1 | 6 - 7 | 1067 | 107 | |
XM_017010814.1 | XP_016866303.1 | 6 - 7 | 1031 | 107 | |
GenBank mRNA | AK125274.1 | - | 6 - 7 | 813 | 107 |
AK298704.1 | - | 6 - 7 | 1067 | 107 | |
AK302420.1 | - | 7 - 8 | 1031 | 107 | |
AK303670.1 | - | 5 - 6 | 1123 | 107 | |
BC009260.2 | - | 6 - 7 | 1093 | 107 |