The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198580.2 | NP_940982.1 | 3 - 4 | 818 | 155 |
XM_011528000.1 | XP_011526302.1 | 4 - 5 | 959 | 155 | |
XM_011528001.2 | XP_011526303.1 | 3 - 4 | 758 | 155 | |
XM_011528002.2 | XP_011526304.1 | 4 - 5 | 739 | 155 | |
XM_011528003.2 | XP_011526305.1 | 4 - 5 | 988 | 155 | |
XM_017026781.1 | XP_016882270.1 | 4 - 5 | 896 | 155 | |
GenBank mRNA | AK294238.1 | - | 3 - 4 | 1180 | 155 |
AK299852.1 | - | 4 - 5 | 1087 | 155 | |
AK315935.1 | - | 2 - 3 | 940 | 155 | |
BC013149.1 | - | 2 - 3 | 308 | 155 | |
BC059399.1 | - | 3 - 4 | 771 | 155 |