The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000068.3 | NP_000059.3 | 11 - 12 | 1798 | 154 |
NM_001127221.1 | NP_001120693.1 | 11 - 12 | 1798 | 154 | |
NM_001127222.1 | NP_001120694.1 | 11 - 12 | 1795 | 154 | |
NM_001174080.1 | NP_001167551.1 | 11 - 12 | 1798 | 154 | |
NM_023035.2 | NP_075461.2 | 11 - 12 | 1798 | 154 | |
GenBank mRNA | AB035726.1 | - | 11 - 12 | 1597 | 154 |
AB035727.2 | - | 11 - 12 | 1559 | 154 | |
AF004883.1 | - | 11 - 12 | 1798 | 154 | |
AF004884.1 | - | 11 - 12 | 1798 | 154 | |
BQ340521.1 | - | 4 - 5 | 426 | 154 | |
FJ040507.1 | - | 11 - 12 | 1562 | 154 | |
U79666.1 | - | 11 - 12 | 1779 | 154 | |
X99897.1 | - | 11 - 12 | 1837 | 154 |