Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CACNA1A
Entrez Gene ID
773
Gene Name
calcium voltage-gated channel subunit alpha1 A
Gene Aliases
APCA, BI, CACNL1A4, CAV2.1, EA2, FHM, HPCA, MHP, MHP1, SCA6
Chromosome Location
Chr.19: 13206442 - 13506460 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000068.3 NP_000059.3 14 - 15 2148 132
NM_001127221.1 NP_001120693.1 14 - 15 2148 132
NM_001127222.1 NP_001120694.1 14 - 15 2145 132
NM_001174080.1 NP_001167551.1 14 - 15 2148 132
NM_023035.2 NP_075461.2 14 - 15 2148 132
GenBank mRNA AB035727.2 - 14 - 15 1909 132
AF004883.1 - 14 - 15 2148 132
AF004884.1 - 14 - 15 2148 132
FJ040507.1 - 14 - 15 1912 132
U79666.1 - 14 - 15 2129 132
X99897.1 - 14 - 15 2187 132

View Full Product Details