The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002897.4 | NP_002888.1 | 14 - 14 | 3167 | 146 |
NM_016836.3 | NP_058520.1 | 14 - 14 | 3176 | 146 | |
XM_005246737.1 | XP_005246794.1 | 15 - 15 | 2935 | 146 | |
XM_005246738.2 | XP_005246795.1 | 15 - 15 | 3270 | 146 | |
XM_005246739.2 | XP_005246796.1 | 15 - 15 | 3140 | 146 | |
XM_005246740.1 | XP_005246797.1 | 15 - 15 | 2926 | 146 | |
XM_005246741.3 | XP_005246798.1 | 15 - 15 | 3137 | 146 | |
XM_017004626.1 | XP_016860115.1 | 14 - 14 | 3213 | 146 | |
GenBank mRNA | AL117617.1 | - | 1 - 1 | 218 | 146 |
BC065192.1 | - | 1 - 1 | 437 | 146 | |
BC069687.1 | - | 1 - 1 | 384 | 146 | |
BC069706.1 | - | 1 - 1 | 383 | 146 | |
BC069763.1 | - | 1 - 1 | 383 | 146 | |
BC080620.1 | - | 1 - 1 | 412 | 147 |