Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MBNL1
Entrez Gene ID
4154
Gene Name
muscleblind like splicing regulator 1
Gene Aliases
EXP, MBNL
Chromosome Location
Chr.3: 152243656 - 152465780 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001314057.1 NP_001300986.1 10 - 11 1776 179
NM_021038.4 NP_066368.2 8 - 9 2507 179
NM_207295.1 NP_997178.1 6 - 7 1678 179
XM_005247457.4 XP_005247514.1 10 - 11 2129 179
XM_005247458.4 XP_005247515.1 10 - 11 3383 179
XM_005247460.4 XP_005247517.1 10 - 11 2092 179
XM_005247462.4 XP_005247519.1 9 - 10 2075 179
XM_005247464.4 XP_005247521.1 9 - 10 2187 179
XM_005247474.3 XP_005247531.1 8 - 9 1047 179
XM_005247475.3 XP_005247532.1 9 - 10 1370 179
XM_005247476.4 XP_005247533.1 7 - 8 1083 179
XM_005247477.4 XP_005247534.1 8 - 9 1136 179
XM_006713639.1 XP_006713702.1 10 - 11 2111 179
XM_011512843.2 XP_011511145.1 9 - 10 1925 179
XM_011512845.2 XP_011511147.1 8 - 9 1871 179
XM_011512847.2 XP_011511149.1 7 - 8 1834 179
XM_011512848.2 XP_011511150.1 10 - 11 1733 179
XM_017006423.1 XP_016861912.1 9 - 10 2380 179
XM_017006424.1 XP_016861913.1 9 - 10 2207 179
XM_017006427.1 XP_016861916.1 8 - 9 2300 179
XM_017006428.1 XP_016861917.1 8 - 9 1964 179
XM_017006429.1 XP_016861918.1 8 - 9 2047 179
XM_017006435.1 XP_016861924.1 9 - 10 1177 179
XM_017006439.1 XP_016861928.1 8 - 9 1316 179
XM_017006440.1 XP_016861929.1 7 - 8 1050 179
GenBank mRNA AF395876.1 - 6 - 7 889 179
AF401998.1 - 7 - 8 1093 179
AJ308400.1 - 7 - 8 1122 179
AK304089.1 - 7 - 8 856 179
BC043493.1 - 8 - 9 2012 179
BC050535.1 - 10 - 11 1297 179

View Full Product Details