Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LDB1
Entrez Gene ID
8861
Gene Name
LIM domain binding 1
Gene Aliases
CLIM-2, CLIM2, LDB-1, NLI
Chromosome Location
Chr.10: 102102100 - 102120473 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113407.2 NP_001106878.1 4 - 5 605 120
NM_001321612.1 NP_001308541.1 4 - 5 605 120
NM_003893.4 NP_003884.1 4 - 5 778 120
XM_017016867.1 XP_016872356.1 4 - 5 313 120
XM_017016868.1 XP_016872357.1 4 - 5 313 120
XM_017016869.1 XP_016872358.1 1 - 2 69 120
GenBank mRNA AB016485.1 - 3 - 4 184 120
AB250384.1 - 3 - 4 141 120
AF064491.1 - 4 - 5 447 120
AF068652.1 - 4 - 5 418 120
AK300588.1 - 4 - 5 645 120
AK308481.1 - 4 - 5 432 120
BC000482.2 - 4 - 5 757 120
BC009246.2 - 4 - 5 757 120
BT007054.1 - 3 - 4 134 120

View Full Product Details