The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001113407.2 | NP_001106878.1 | 9 - 10 | 1220 | 102 |
NM_001321612.1 | NP_001308541.1 | 9 - 10 | 1220 | 102 | |
NM_003893.4 | NP_003884.1 | 9 - 10 | 1393 | 102 | |
XM_017016867.1 | XP_016872356.1 | 9 - 10 | 928 | 102 | |
XM_017016868.1 | XP_016872357.1 | 9 - 10 | 928 | 102 | |
XM_017016869.1 | XP_016872358.1 | 5 - 6 | 511 | 102 | |
GenBank mRNA | AB016485.1 | - | 8 - 9 | 799 | 102 |
AB250384.1 | - | 8 - 9 | 756 | 102 | |
AF064491.1 | - | 9 - 10 | 1062 | 102 | |
AF068652.1 | - | 9 - 10 | 1033 | 102 | |
AK300588.1 | - | 9 - 10 | 1260 | 102 | |
AK308481.1 | - | 9 - 10 | 1047 | 102 | |
BC000482.2 | - | 9 - 10 | 1372 | 102 | |
BC009246.2 | - | 9 - 10 | 1372 | 102 | |
BT007054.1 | - | 8 - 9 | 749 | 102 |