Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCNT2
Entrez Gene ID
343450
Gene Name
potassium sodium-activated channel subfamily T member 2
Gene Aliases
KCa4.2, SLICK, SLO2.1
Chromosome Location
Chr.1: 196225779 - 196608905 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001287819.1 NP_001274748.1 17 - 18 2118 147
NM_001287820.1 NP_001274749.1 16 - 17 1968 147
NM_198503.3 NP_940905.2 17 - 18 2118 147
XM_006711294.2 XP_006711357.1 17 - 18 2592 147
XM_006711295.2 XP_006711358.1 17 - 18 2592 147
XM_011509482.2 XP_011507784.1 16 - 17 2517 147
XM_011509483.2 XP_011507785.1 17 - 18 2592 147
XM_017001178.1 XP_016856667.1 17 - 18 2592 147
XM_017001179.1 XP_016856668.1 17 - 18 2592 147
XM_017001180.1 XP_016856669.1 17 - 18 2592 147
XM_017001181.1 XP_016856670.1 7 - 8 1107 147
XM_017001182.1 XP_016856671.1 16 - 17 2483 147
XM_017001183.1 XP_016856672.1 4 - 5 643 147
XM_017001184.1 XP_016856673.1 16 - 17 2483 147
XM_017001185.1 XP_016856674.1 16 - 17 2517 147
GenBank mRNA AK127807.1 - 10 - 11 1468 147
AK131498.1 - 16 - 17 2193 147
AK309729.1 - 17 - 18 2107 147
AY359444.1 - 17 - 18 2056 147
BC103947.1 - 13 - 14 1499 147
BC103948.1 - 17 - 18 2046 147
BC103949.1 - 16 - 17 1896 147
BC103950.1 - 17 - 18 2046 147
BX647852.1 - 17 - 18 2100 147
EU251076.1 - 17 - 18 1996 147

View Full Product Details