The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001287819.1 | NP_001274748.1 | 19 - 20 | 2399 | 130 |
NM_001287820.1 | NP_001274749.1 | 18 - 19 | 2249 | 130 | |
XM_006711295.2 | XP_006711358.1 | 19 - 20 | 2873 | 130 | |
XM_017001178.1 | XP_016856667.1 | 19 - 20 | 2873 | 130 | |
XM_017001185.1 | XP_016856674.1 | 18 - 19 | 2798 | 130 | |
GenBank mRNA | AK131498.1 | - | 18 - 19 | 2474 | 130 |
AK309729.1 | - | 19 - 20 | 2388 | 130 | |
BC103948.1 | - | 19 - 20 | 2327 | 130 | |
BC103949.1 | - | 18 - 19 | 2177 | 130 | |
BC103950.1 | - | 19 - 20 | 2327 | 130 | |
BX647852.1 | - | 19 - 20 | 2381 | 130 |