Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCNT2
Entrez Gene ID
343450
Gene Name
potassium sodium-activated channel subfamily T member 2
Gene Aliases
KCa4.2, SLICK, SLO2.1
Chromosome Location
Chr.1: 196225779 - 196608905 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001287819.1 NP_001274748.1 20 - 21 2531 142
NM_001287820.1 NP_001274749.1 19 - 20 2381 142
NM_198503.3 NP_940905.2 21 - 22 2603 142
XM_006711294.2 XP_006711357.1 21 - 22 3077 142
XM_006711295.2 XP_006711358.1 20 - 21 3005 142
XM_011509482.2 XP_011507784.1 20 - 21 3002 142
XM_017001178.1 XP_016856667.1 20 - 21 3005 142
XM_017001179.1 XP_016856668.1 21 - 22 3077 142
XM_017001181.1 XP_016856670.1 11 - 12 1592 142
XM_017001182.1 XP_016856671.1 20 - 21 2968 142
XM_017001183.1 XP_016856672.1 8 - 9 1128 142
XM_017001184.1 XP_016856673.1 20 - 21 2968 142
XM_017001185.1 XP_016856674.1 19 - 20 2930 142
GenBank mRNA AK127807.1 - 13 - 14 1804 142
AK131498.1 - 19 - 20 2606 142
AK309729.1 - 20 - 21 2520 142
AY359444.1 - 21 - 22 2541 142
BC103948.1 - 20 - 21 2459 142
BC103949.1 - 19 - 20 2309 142
BC103950.1 - 20 - 21 2459 142
BX647852.1 - 20 - 21 2513 142
EU251076.1 - 21 - 22 2481 142

View Full Product Details