Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCNT2
Entrez Gene ID
343450
Gene Name
potassium sodium-activated channel subfamily T member 2
Gene Aliases
KCa4.2, SLICK, SLO2.1
Chromosome Location
Chr.1: 196225779 - 196608905 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001287819.1 NP_001274748.1 23 - 24 2828 145
NM_001287820.1 NP_001274749.1 22 - 23 2678 145
NM_198503.3 NP_940905.2 24 - 25 2900 145
XM_006711294.2 XP_006711357.1 24 - 25 3374 145
XM_006711295.2 XP_006711358.1 23 - 24 3302 145
XM_011509482.2 XP_011507784.1 23 - 24 3299 145
XM_017001178.1 XP_016856667.1 23 - 24 3302 145
XM_017001181.1 XP_016856670.1 14 - 15 1889 145
XM_017001182.1 XP_016856671.1 23 - 24 3265 145
XM_017001183.1 XP_016856672.1 11 - 12 1425 145
XM_017001184.1 XP_016856673.1 23 - 24 3265 145
XM_017001185.1 XP_016856674.1 22 - 23 3227 145
GenBank mRNA AK127807.1 - 16 - 17 2101 145
AK131498.1 - 22 - 23 2903 145
AK309729.1 - 23 - 24 2817 145
AY359444.1 - 24 - 25 2838 145
BC103947.1 - 18 - 19 2092 145
BC103948.1 - 23 - 24 2756 145
BC103949.1 - 22 - 23 2606 145
BC103950.1 - 23 - 24 2756 145
BX647852.1 - 23 - 24 2810 145
EU251076.1 - 24 - 25 2778 145

View Full Product Details