Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCNT2
Entrez Gene ID
343450
Gene Name
potassium sodium-activated channel subfamily T member 2
Gene Aliases
KCa4.2, SLICK, SLO2.1
Chromosome Location
Chr.1: 196225779 - 196608905 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001287819.1 NP_001274748.1 7 - 8 671 154
NM_001287820.1 NP_001274749.1 7 - 8 671 154
NM_198503.3 NP_940905.2 7 - 8 671 154
XM_006711294.2 XP_006711357.1 7 - 8 1145 154
XM_006711295.2 XP_006711358.1 7 - 8 1145 154
XM_011509482.2 XP_011507784.1 6 - 7 1070 154
XM_011509483.2 XP_011507785.1 7 - 8 1145 154
XM_017001178.1 XP_016856667.1 7 - 8 1145 154
XM_017001179.1 XP_016856668.1 7 - 8 1145 154
XM_017001180.1 XP_016856669.1 7 - 8 1145 154
XM_017001182.1 XP_016856671.1 7 - 8 1145 154
XM_017001184.1 XP_016856673.1 7 - 8 1145 154
XM_017001185.1 XP_016856674.1 6 - 7 1070 154
GenBank mRNA AK131498.1 - 6 - 7 725 154
AK309729.1 - 7 - 8 660 154
AY359444.1 - 7 - 8 609 154
BC103948.1 - 7 - 8 599 154
BC103949.1 - 7 - 8 599 154
BC103950.1 - 7 - 8 599 154
BX647852.1 - 7 - 8 652 154
EU251076.1 - 7 - 8 549 154

View Full Product Details