The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_207662.3 | NP_997545.2 | 1 - 2 | 719 | 105 |
XM_005268073.3 | XP_005268130.1 | 1 - 2 | 719 | 105 | |
GenBank mRNA | BC027607.2 | - | 1 - 2 | 705 | 105 |