Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UPK3B
Entrez Gene ID
80761
Gene Name
uroplakin 3B
Gene Aliases
P35, UP3B, UPIIIB
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030570.3 NP_085047.1 3 - 4 984 112
NM_182683.2 NP_872624.1 4 - 5 819 112
NM_182684.2 NP_872625.1 5 - 6 899 112
GenBank mRNA AK290096.1 - 3 - 4 887 112
AY233462.1 - 5 - 6 732 112
BC004304.1 - 3 - 4 808 112

Gene Symbol
LOC105375355
Entrez Gene ID
105375355
Gene Name
uroplakin-3b
Gene Aliases
-
Chromosome Location
Chr.7: 76510102 - 76524676 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005250612.3 XP_005250669.1 3 - 4 1305 112
XM_006716140.3 XP_006716203.1 5 - 6 977 112
XM_006716141.3 XP_006716204.1 4 - 5 897 112

View Full Product Details