Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HEATR4
Entrez Gene ID
399671
Gene Name
HEAT repeat containing 4
Gene Aliases
-
Chromosome Location
Chr.14: 73478484 - 73634418 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001220484.1 NP_001207413.1 3 - 4 1227 122
NM_203309.2 NP_976054.2 2 - 3 1148 122
XM_006720143.2 XP_006720206.1 2 - 3 1185 122
XM_011536760.2 XP_011535062.1 2 - 3 1371 122
XM_011536761.2 XP_011535063.1 2 - 3 1030 122
XM_017021289.1 XP_016876778.1 2 - 3 1709 122
GenBank mRNA AK093515.1 - 2 - 3 1141 122
AK302564.1 - 3 - 4 1201 122
BC047590.1 - 2 - 3 1148 122
GU727638.1 - 1 - 2 738 122

View Full Product Details