Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HEATR4
Entrez Gene ID
399671
Gene Name
HEAT repeat containing 4
Gene Aliases
-
Chromosome Location
Chr.14: 73478484 - 73634418 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001220484.1 NP_001207413.1 4 - 5 1411 119
NM_203309.2 NP_976054.2 3 - 4 1332 119
XM_006720143.2 XP_006720206.1 3 - 4 1369 119
XM_011536760.2 XP_011535062.1 3 - 4 1555 119
XM_011536761.2 XP_011535063.1 3 - 4 1214 119
XM_017021289.1 XP_016876778.1 3 - 4 1893 119
GenBank mRNA AK093515.1 - 3 - 4 1325 119
AK302564.1 - 4 - 5 1385 119
BC047590.1 - 3 - 4 1332 119
GU727638.1 - 2 - 3 922 119

View Full Product Details