Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HEATR4
Entrez Gene ID
399671
Gene Name
HEAT repeat containing 4
Gene Aliases
-
Chromosome Location
Chr.14: 73478484 - 73634418 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001220484.1 NP_001207413.1 7 - 8 1909 149
NM_203309.2 NP_976054.2 6 - 7 1830 149
XM_006720143.2 XP_006720206.1 6 - 7 1867 149
XM_011536760.2 XP_011535062.1 6 - 7 2053 149
XM_011536761.2 XP_011535063.1 6 - 7 1712 149
XM_017021289.1 XP_016876778.1 6 - 7 2391 149
GenBank mRNA AK302564.1 - 7 - 8 1883 149
BC047590.1 - 6 - 7 1830 149
GU727638.1 - 5 - 6 1420 149

View Full Product Details