Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HEATR4
Entrez Gene ID
399671
Gene Name
HEAT repeat containing 4
Gene Aliases
-
Chromosome Location
Chr.14: 73478484 - 73634418 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001220484.1 NP_001207413.1 11 - 12 2449 166
NM_203309.2 NP_976054.2 10 - 11 2370 166
XM_006720143.2 XP_006720206.1 10 - 11 2407 166
XM_011536760.2 XP_011535062.1 10 - 11 2593 166
XM_017021289.1 XP_016876778.1 10 - 11 2931 166
GenBank mRNA AK302564.1 - 11 - 12 2423 166
BC047590.1 - 10 - 11 166
GU727638.1 - 9 - 10 1960 166

View Full Product Details