The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152903.4 | NP_690867.3 | 1 - 1 | 2729 | 150 |
GenBank mRNA | AK095195.1 | - | 1 - 1 | 1374 | 150 |
AK096608.1 | - | 2 - 2 | 2669 | 150 | |
AL833839.1 | - | 1 - 1 | 2381 | 150 | |
BC000560.2 | - | 1 - 1 | 2651 | 150 | |
BC051349.1 | - | 1 - 1 | 2490 | 150 |