Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ITGB1BP1
Entrez Gene ID
9270
Gene Name
integrin subunit beta 1 binding protein 1
Gene Aliases
ICAP-1A, ICAP-1B, ICAP-1alpha, ICAP1, ICAP1A, ICAP1B
Chromosome Location
Chr.2: 9405681 - 9423569 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001319068.1 NP_001305997.1 1 - 2 182 159
XM_017005269.1 XP_016860758.1 1 - 2 738 159
GenBank mRNA AK093179.1 - 1 - 2 771 159
BE146261.1 - 2 - 3 159
BU520903.1 - 1 - 2 156 159

View Full Product Details