Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ITGB1BP1
Entrez Gene ID
9270
Gene Name
integrin subunit beta 1 binding protein 1
Gene Aliases
ICAP-1A, ICAP-1B, ICAP-1alpha, ICAP1, ICAP1A, ICAP1B
Chromosome Location
Chr.2: 9405681 - 9423569 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005246183.4 XP_005246240.1 2 - 3 884 142
XM_005246184.4 XP_005246241.1 2 - 3 323 142
XM_005246185.4 XP_005246242.1 2 - 3 196 142
XM_005246189.4 XP_005246246.1 2 - 3 849 142
XM_011510416.2 XP_011508718.1 2 - 3 518 142
XM_017005268.1 XP_016860757.1 2 - 3 188 142

View Full Product Details