The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001285444.1 | NP_001272373.1 | 18 - 19 | 1964 | 153 |
NM_001285445.1 | NP_001272374.1 | 18 - 19 | 1961 | 153 | |
NM_001285447.1 | NP_001272376.1 | 19 - 20 | 1857 | 153 | |
NM_001285448.1 | NP_001272377.1 | 18 - 19 | 1870 | 153 | |
NM_001324019.1 | NP_001310948.1 | 19 - 20 | 2042 | 153 | |
NM_015027.3 | NP_055842.2 | 19 - 20 | 2045 | 153 | |
XM_005255173.1 | XP_005255230.1 | 19 - 20 | 2001 | 153 | |
XM_005255176.2 | XP_005255233.1 | 19 - 20 | 1927 | 153 | |
XM_006720865.2 | XP_006720928.2 | 18 - 19 | 1763 | 153 | |
XM_017023059.1 | XP_016878548.1 | 20 - 21 | 1937 | 153 | |
XM_017023060.1 | XP_016878549.1 | 19 - 20 | 2107 | 153 | |
XM_017023061.1 | XP_016878550.1 | 19 - 20 | 1877 | 153 | |
XM_017023062.1 | XP_016878551.1 | 19 - 20 | 1958 | 153 | |
XM_017023063.1 | XP_016878552.1 | 20 - 21 | 1937 | 153 | |
XM_017023064.1 | XP_016878553.1 | 18 - 19 | 1865 | 153 | |
GenBank mRNA | AK025504.1 | - | 6 - 7 | 679 | 153 |
AK295168.1 | - | 19 - 20 | 1850 | 153 | |
AK299111.1 | - | 18 - 19 | 1795 | 153 | |
AK299799.1 | - | 18 - 19 | 1855 | 153 | |
BC025366.2 | - | 13 - 14 | 1231 | 153 | |
BC036520.1 | - | 18 - 19 | 2863 | 153 | |
BC042104.2 | - | 2 - 3 | 144 | 153 | |
BC060871.1 | - | 19 - 20 | 1879 | 153 | |
BX648066.1 | - | 18 - 19 | 1802 | 153 | |
D87438.1 | - | 19 - 20 | 1905 | 153 |