Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLPPR4
Entrez Gene ID
9890
Gene Name
phospholipid phosphatase related 4
Gene Aliases
LPPR4, LPR4, PHP1, PRG-1, PRG1
Chromosome Location
Chr.1: 99262956 - 99309584 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166252.1 NP_001159724.1 3 - 4 696 107
NM_014839.4 NP_055654.2 3 - 4 696 107
XM_011542498.2 XP_011540800.1 3 - 4 644 107
GenBank mRNA AB007924.3 - 3 - 4 692 107
AF357013.1 - 3 - 4 469 107
AF541281.1 - 3 - 4 703 107
AK291369.1 - 3 - 4 496 107
AK300361.1 - 3 - 4 440 107
AL834390.1 - 2 - 3 209 107
AY304518.1 - 3 - 4 644 107
BC042963.1 - 3 - 4 514 107

View Full Product Details