The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012191.3 | NP_036323.2 | 1 - 2 | 212 | 132 |
GenBank mRNA | BC004483.2 | - | 1 - 2 | 72 | 132 |