Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SEC14L4
Entrez Gene ID
284904
Gene Name
SEC14 like lipid binding 4
Gene Aliases
TAP3
Chromosome Location
Chr.22: 30488150 - 30505711 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161368.1 NP_001154840.1 4 - 5 338 122
NM_174977.3 NP_777637.1 4 - 5 338 122
XM_006724235.2 XP_006724298.1 4 - 5 354 122
XM_011530164.2 XP_011528466.1 4 - 5 518 122
XM_017028787.1 XP_016884276.1 3 - 4 467 122
XM_017028788.1 XP_016884277.1 3 - 4 468 122
XM_017028789.1 XP_016884278.1 3 - 4 140 122
XM_017028790.1 XP_016884279.1 3 - 4 475 122
XM_017028791.1 XP_016884280.1 3 - 4 167 122
GenBank mRNA AK092002.1 - 5 - 6 387 122
AK093418.1 - 4 - 5 355 122
AK093448.1 - 4 - 5 322 122
AY158085.1 - 4 - 5 238 122
BC136358.1 - 4 - 5 310 122
BC136359.1 - 4 - 5 310 122
BC139912.1 - 4 - 5 277 122
CR456346.1 - 2 - 3 97 122

View Full Product Details