Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CIDEB
Entrez Gene ID
27141
Gene Name
cell death-inducing DFFA-like effector b
Gene Aliases
-
Chromosome Location
Chr.14: 24305187 - 24311377 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001318807.1 NP_001305736.1 2 - 3 872 179

View Full Product Details