The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178821.1 | NP_849143.1 | 4 - 5 | 395 | 165 |
XM_011510755.1 | XP_011509057.1 | 5 - 6 | 1004 | 165 | |
XM_017003490.1 | XP_016858979.1 | 3 - 4 | 410 | 165 | |
GenBank mRNA | AK097409.1 | - | 4 - 5 | 395 | 165 |
AK098821.1 | - | 4 - 5 | 343 | 165 | |
AK127882.1 | - | 4 - 5 | 952 | 165 | |
AK301861.1 | - | 5 - 6 | 395 | 165 | |
BC036377.2 | - | 4 - 5 | 375 | 165 |