The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178821.1 | NP_849143.1 | 8 - 9 | 836 | 145 |
XM_011510755.1 | XP_011509057.1 | 9 - 10 | 1445 | 145 | |
XM_017003490.1 | XP_016858979.1 | 7 - 8 | 851 | 145 | |
GenBank mRNA | AK097409.1 | - | 8 - 9 | 836 | 145 |
AK098821.1 | - | 8 - 9 | 784 | 145 | |
AK127882.1 | - | 8 - 9 | 1393 | 145 | |
AK301861.1 | - | 9 - 10 | 836 | 145 | |
BC036377.2 | - | 8 - 9 | 816 | 145 |